2-Chloro-4,5-difluorophenol ≥97%

Supplier: AOBChem USA

Warning

36948-1G 36948-25G 36948-500MG 36948-5G
76903-374EA 296 USD
76903-374 76903-376 76903-378 76903-380
2-Chloro-4,5-difluorophenol ≥97%
2-Chloro-4,5-difluorophenol

Precaución: for Research Use Only

Formula: C₆H₃ClF₂O
MW: 164.537 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18415628
CAS Number: 2267-98-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR