4-(2-Methylphenyl)benzonitrile ≥95%

Supplier: AOBChem USA

Warning

36772-1G 36772-25G 36772-500MG 36772-5G
76903-362EA 142.17 USD
76903-362 76903-364 76903-366 76903-368
4-(2-Methylphenyl)benzonitrile ≥95%
4-(2-Methylphenyl)benzonitrile

Precaución: for Research Use Only

Formula: C₁₄H₁₁N
Storage Temperature: Refrigerator
MDL Number: MFCD05981801
CAS Number: 189828-30-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR