2-Ethoxy-4-fluoro-1-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

36604-25G 36604-5G
76903-320EA 481.65 USD
76903-320 76903-322
2-Ethoxy-4-fluoro-1-methylbenzene ≥97%
2-Ethoxy-4-fluoro-1-methylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₁FO
MW: 154.181 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32632045
CAS Number: 1691815-54-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR