2-Methyl-3-(methylthio)benzoic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
36552-1G
36552-25G
36552-500MG
36552-5G
76903-308EA
421.18
USD
76903-308
76903-310
76903-312
76903-314
2-Methyl-3-(methylthio)benzoic acid ≥97%
2-Methyl-3-(methylthio)benzoic acid
Precaución: for Research Use Only
Formula:
C₉H₁₀O₂S Storage Temperature: Ambient |
MDL Number:
MFCD20660289 CAS Number: 1149384-42-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...