2,6-Dibromo-4-iodophenol ≥95%

Supplier: AOBChem USA

Warning

36524-1G 36524-25G 36524-500MG 36524-5G
76903-298EA 265.23 USD
76903-298 76903-300 76903-302 76903-304
2,6-Dibromo-4-iodophenol ≥95%
2,6-Dibromo-4-iodophenol

Precaución: for Research Use Only

Formula: C₆H₃Br₂IO
Storage Temperature: Refrigerator
MDL Number: MFCD20923585
CAS Number: 187407-15-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR