2-Chloro-4-phenoxyphenol ≥97%

Supplier: AOBChem USA

Warning

35793-1G 35793-25G 35793-500MG 35793-5G
76903-198EA 237.65 USD
76903-198 76903-200 76903-202 76903-204
2-Chloro-4-phenoxyphenol ≥97%
2-Chloro-4-phenoxyphenol

Precaución: for Research Use Only

Formula: C₁₂H₉ClO₂
MW: 220.652 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD27925180
CAS Number: 54582-59-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR