2,5-Dimethoxyterephthalaldehyde ≥95%

Supplier: AOBChem USA

Warning

35752-1G 35752-25G 35752-5G
76903-188EA 99.73 USD
76903-188 76903-190 76903-192
2,5-Dimethoxyterephthalaldehyde ≥95%
2,5-Dimethoxyterephthalaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₄
Storage Temperature: Ambient
MDL Number: MFCD00017606
CAS Number: 7310-97-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR