2-Bromo-6-chloroisonicotinic acid ≥95%

Supplier: AOBChem USA

Warning

35614-1G 35614-25G 35614-500MG 35614-5G
76903-170EA 350.1 USD
76903-170 76903-172 76903-174 76903-176
2-Bromo-6-chloroisonicotinic acid ≥95%
2-Bromo-6-chloroisonicotinic acid

Precaución: for Research Use Only

Formula: C₆H₃BrClNO₂
MW: 236.451 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13185792
CAS Number: 1060811-26-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR