2-Chloro-3-methoxy-4-methylpyridine ≥97%
Supplier: AOBChem USA
|
Warning
|
35554-1G
35554-25G
35554-500MG
35554-5G
76903-154EA
441.34
USD
76903-154
76903-156
76903-158
76903-160
2-Chloro-3-methoxy-4-methylpyridine ≥97%
2-Chloro-3-methoxy-4-methylpyridine
Precaución: for Research Use Only
Formula:
C₇H₈ClNO Storage Temperature: Ambient |
MDL Number:
MFCD16610588 CAS Number: 1227562-13-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...