2-Chloro-3-(Methylsulfanyl)Benzoic Acid ≥97%

Supplier: AOBChem USA

Warning

35283-1G 35283-500MG 35283-5G
76903-098EA 558.04 USD
76903-098 76903-100 76903-102
2-Chloro-3-(Methylsulfanyl)Benzoic Acid ≥97%
2-Chloro-3-(Methylsulfanyl)Benzoic Acid

Precaución: for Research Use Only

Formula: C₈H₇ClO₂S
Storage Temperature: Refrigerator
MDL Number: MFCD22489493
CAS Number: 1805648-85-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR