2-Fluoro-6-methylphenol 95% research grade

Supplier: AOBChem USA

Warning

Synonyms: 3-Fluoro-2-hydroxytoluene

35213-1G 35213-25G 35213-5G 35213-500MG
76903-086EA 106.09 USD
76903-086 76903-088 76903-090 77646-280
2-Fluoro-6-methylphenol 95% research grade
2-Fluoro-6-methylphenol
Formula: C₇H₇FO
Storage Temperature: Refrigerator
MDL Number: MFCD12026463
CAS Number: 443-90-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR