2-Bromo-4-fluoro-3-iodobenzotrifluoride ≥95%

Supplier: AOBChem USA

Warning

34893-1G 34893-25G 34893-500MG 34893-5G
76903-054EA 350.1 USD
76903-054 76903-056 76903-058 76903-060
2-Bromo-4-fluoro-3-iodobenzotrifluoride ≥95%
2-Bromo-4-fluoro-3-iodobenzotrifluoride
Formula: C₇H₂BrF₄I
Storage Temperature: Ambient
MDL Number: MFCD31568566
CAS Number: 2227272-51-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR