3-(2-Methoxyphenyl)pyridine ≥95%

Supplier: AOBChem USA

Warning

34891-1G 34891-25G 34891-500MG 34891-5G
76903-040EA 269.47 USD
76903-040 76903-042 76903-044 76903-046
3-(2-Methoxyphenyl)pyridine ≥95%
3-(2-Methoxyphenyl)pyridine

Precaución: for Research Use Only

Formula: C₁₂H₁₁NO
Storage Temperature: Refrigerator
MDL Number: MFCD06801827
CAS Number: 5958-01-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR