2,6-Difluoro-3-hydroxybenzonitrile ≥97%

Supplier: AOBChem USA

Warning

34849-1G 34849-500MG 34849-5G
76903-026EA 468.92 USD
76903-026 76903-028 76903-030
2,6-Difluoro-3-hydroxybenzonitrile ≥97%
2,6-Difluoro-3-hydroxybenzonitrile

Precaución: for Research Use Only

Formula: C₇H₃F₂NO
MW: 155.102 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09163028
CAS Number: 946796-26-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR