2-Chloro-5-(methylthio)benzotrifluoride ≥97%

Supplier: AOBChem USA

Warning

34776-1G 34776-25G 34776-500MG 34776-5G
76902-990EA 290.69 USD
76902-990 76902-992 76902-994 76902-996
2-Chloro-5-(methylthio)benzotrifluoride ≥97%
2-Chloro-5-(methylthio)benzotrifluoride

Precaución: for Research Use Only

Formula: C₈H₆ClF₃S
Storage Temperature: Refrigerator
MDL Number: MFCD09878422
CAS Number: 957066-04-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR