2-Methyl-5-propoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

34760-1G 34760-500MG 34760-5G
76902-980EA 524.09 USD
76902-980 76902-982 76902-984
2-Methyl-5-propoxybenzoic acid ≥97%
2-Methyl-5-propoxybenzoic acid
Formula: C₁₁H₁₄O₃
MW: 194.227 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19252307
CAS Number: 1282679-69-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR