2-Ethoxy-5-fluorobenzoic acid ≥97% research grade

Supplier: AOBChem USA

Warning

34744-1G 34744-25G 34744-500MG 34744-5G
76902-964EA 289.63 USD
76902-964 76902-966 76902-968 76902-970
2-Ethoxy-5-fluorobenzoic acid ≥97% research grade
2-Ethoxy-5-fluorobenzoic acid
Formula: C₉H₉FO₃
Storage Temperature: Refrigerator
MDL Number: MFCD11639882
CAS Number: 445-93-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR