2'-Bromo-[1,1'-biphenyl]-4-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

34741-1G 34741-500MG 34741-5G
76902-950EA 551.67 USD
76902-950 76902-952 76902-954
2'-Bromo-[1,1'-biphenyl]-4-carbaldehyde ≥97%
2'-Bromo-[1,1'-biphenyl]-4-carbaldehyde
Formula: C₁₃H₉BrO
MW: 261.114 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD05980885
CAS Number: 198205-81-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR