2-Iodo-4-(trifluoromethyl)anisole ≥95%

Supplier: AOBChem USA

Warning

34652-1G 34652-25G 34652-500MG 34652-5G
76902-934EA 135.8 USD
76902-934 76902-936 76902-938 76902-940
2-Iodo-4-(trifluoromethyl)anisole ≥95%
2-Iodo-4-(trifluoromethyl)anisole

Precaución: for Research Use Only

Formula: C₈H₆F₃IO
MW: 302.03 g/mol
Storage Temperature: Ambient
CAS Number: 195624-84-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR