2-(2-Bromo-5-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Bromo-5-methoxyphenylboronic acid pinacol ester

34627-1G 34627-25G 34627-5G
76902-922EA 62.6 USD
76902-922 76902-924 76902-926
2-(2-Bromo-5-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%
2-(2-Bromo-5-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₃H₁₈BBrO₃
MW: 312.995 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17215810
CAS Number: 1256781-58-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR