2-Chloro-3-fluoro-6-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

34379-1G 34379-25G 34379-500MG 34379-5G
76902-858EA 222.79 USD
76902-858 76902-860 76902-862 76902-864
2-Chloro-3-fluoro-6-methoxybenzaldehyde ≥97%
2-Chloro-3-fluoro-6-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆ClFO₂
MW: 188.583 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18432476
CAS Number: 1263378-40-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR