2-Bromo-6-iodo-4-(trifluoromethyl)aniline ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Amino-3-bromo-5-iodobenzotrifluoride

34188-1G 34188-25G 34188-5G
76902-822EA 82.76 USD
76902-822 76902-824 76902-826
2-Bromo-6-iodo-4-(trifluoromethyl)aniline ≥97%
2-Bromo-6-iodo-4-(trifluoromethyl)aniline

Precaución: for Research Use Only

Formula: C₇H₄BrF₃IN
MW: 365.9170 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08458149
CAS Number: 875306-20-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR