2-Chloro-5-formylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

34057-1G 34057-500MG 34057-5G
76902-794EA 587.74 USD
76902-794 76902-796 76902-798
2-Chloro-5-formylbenzoic acid ≥95%
2-Chloro-5-formylbenzoic acid
Formula: C₈H₅ClO₃
MW: 184.577 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18073622
CAS Number: 1206625-81-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR