2-Chloro-3-fluoro-5-methylphenol ≥97% research grade

Supplier: AOBChem USA

Warning

34051-1G 34051-500MG 34051-5G
76902-782EA 706.56 USD
76902-782 76902-784 76902-786
2-Chloro-3-fluoro-5-methylphenol ≥97% research grade
2-Chloro-3-fluoro-5-methylphenol
Formula: C₇H₆ClFO
Storage Temperature: Refrigerator
MDL Number: MFCD28752132
CAS Number: 1807168-54-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR