2-Chloro-3,6-difluorophenol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Choro-3,6-difluorophenol

34021-25G 34021-5G
76902-776EA 381.93 USD
76902-776 76902-778
2-Chloro-3,6-difluorophenol ≥97%
2-Chloro-3,6-difluorophenol

Precaución: for Research Use Only

Formula: C₆H₃ClF₂O
MW: 164.54 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01631450
CAS Number: 261762-50-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR