2-Chloro-5-isopropoxyphenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

33825-1G 33825-25G 33825-5G
76902-748EA 96.55 USD
76902-748 76902-750 76902-752
2-Chloro-5-isopropoxyphenylboronic acid ≥95%
2-Chloro-5-isopropoxyphenylboronic acid
Formula: C₉H₁₂BClO₃
Storage Temperature: Refrigerator
MDL Number: MFCD16295073
CAS Number: 1256346-12-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR