2-Chloro-5-cyanobenzoic acid ≥95%

Supplier: AOBChem USA

Warning

33780-1G 33780-25G 33780-500MG 33780-5G
76902-734EA 267.35 USD
76902-734 76902-736 76902-738 76902-740
2-Chloro-5-cyanobenzoic acid ≥95%
2-Chloro-5-cyanobenzoic acid

Precaución: for Research Use Only

Formula: C₈H₄ClNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD11036379
CAS Number: 89891-83-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR