2,4-Difluoro-3-cyanophenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

33758-1G 33758-25G 33758-5G
76902-720EA 107.16 USD
76902-720 76902-722 76902-724
2,4-Difluoro-3-cyanophenylboronic acid pinacol ester ≥97%
2,4-Difluoro-3-cyanophenylboronic acid pinacol ester
Formula: C₁₃H₁₄BF₂NO₂
MW: 265.064 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD31561325
CAS Number: 2216724-47-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR