2-Methylthiophene-3-carboxylic acid ≥97% research grade

Supplier: AOBChem USA

Warning

33721-25G 33721-5G
76902-700EA 187.78 USD
76902-700 76902-702
2-Methylthiophene-3-carboxylic acid ≥97% research grade
2-Methylthiophene-3-carboxylic acid
Formula: C₆H₆O₂S
Storage Temperature: Ambient
MDL Number: MFCD01859891
CAS Number: 1918-78-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR