2-Bromo-3,6-difluorobenzyl alcohol ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
(2-Bromo-3,6-difluorophenyl)methanol
33711-1G
33711-25G
33711-500MG
33711-5G
76902-684EA
231.28
USD
76902-684
76902-686
76902-688
76902-690
2-Bromo-3,6-difluorobenzyl alcohol ≥97%
2-Bromo-3,6-difluorobenzyl alcohol
Precaución: for Research Use Only
Formula:
C₇H₅BrF₂O MW: 223.015 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD28061449 CAS Number: 1404115-38-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...