3-Fluoro-2,4-dimethylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

33641-1G 33641-25G 33641-500MG 33641-5G
76902-652EA 186.72 USD
76902-652 76902-654 76902-656 76902-658
3-Fluoro-2,4-dimethylbenzaldehyde ≥97%
3-Fluoro-2,4-dimethylbenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉FO
Storage Temperature: Refrigerator
MDL Number: MFCD04972838
CAS Number: 363134-36-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR