2,5-Dichloro-3-methoxybenzoic acid ≥97% research grade

Supplier: AOBChem USA

Warning

33592-1G 33592-25G 33592-500MG 33592-5G
76902-636EA 290.69 USD
76902-636 76902-638 76902-640 76902-642
2,5-Dichloro-3-methoxybenzoic acid ≥97% research grade
2,5-Dichloro-3-methoxybenzoic acid
Formula: C₈H₆Cl₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD31382558
CAS Number: 33234-25-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR