2,5-Dichloro-3-fluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

33567-1G 33567-25G 33567-500MG 33567-5G
76902-608EA 264.17 USD
76902-608 76902-610 76902-612 76902-614
2,5-Dichloro-3-fluorophenylboronic acid ≥97%
2,5-Dichloro-3-fluorophenylboronic acid
Formula: C₆H₄BCl₂FO₂
MW: 208.81 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22414432
CAS Number: 2121511-41-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR