2,6-Difluoro-4-pyridinemethanol ≥97% research grade

Supplier: AOBChem USA

Warning

33475-1G 33475-500MG 33475-5G
76902-564EA 560.16 USD
76902-564 76902-566 76902-568
2,6-Difluoro-4-pyridinemethanol ≥97% research grade
2,6-Difluoro-4-pyridinemethanol
Formula: C₆H₅F₂NO
Storage Temperature: Refrigerator
MDL Number: MFCD16250689
CAS Number: 1006299-98-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR