2-Chloro-3-(methoxymethoxy)pyridine ≥97%

Supplier: AOBChem USA

Warning

33369-1G 33369-25G 33369-5G
76902-552EA 96.55 USD
76902-552 76902-554 76902-556
2-Chloro-3-(methoxymethoxy)pyridine ≥97%
2-Chloro-3-(methoxymethoxy)pyridine
Formula: C₇H₈ClNO₂
MW: 173.5969 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD21195353
CAS Number: 862667-72-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR