2-Fluoro-5-(propan-2-yloxy)phenol ≥97% research grade

Supplier: AOBChem USA

Warning

33292-1G 33292-25G 33292-500MG 33292-5G
76902-538EA 159.14 USD
76902-538 76902-540 76902-542 76902-544
2-Fluoro-5-(propan-2-yloxy)phenol ≥97% research grade
2-Fluoro-5-(propan-2-yloxy)phenol
Formula: C₉H₁₁FO₂
MW: 170.181 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16997357
CAS Number: 1243289-64-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR