2-Methoxy-4,6-bis(trifluoromethyl)phenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

33254-1G 33254-25G 33254-500MG 33254-5G
76902-522EA 150.65 USD
76902-522 76902-524 76902-526 76902-528
2-Methoxy-4,6-bis(trifluoromethyl)phenylboronic acid pinacol ester ≥97%
2-Methoxy-4,6-bis(trifluoromethyl)phenylboronic acid pinacol ester
Formula: C₁₅H₁₇BF₆O₃
MW: 370.095 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD30737498
CAS Number: 2121512-96-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR