2-Chloro-1-(methoxymethoxy)-3(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

33222-25G 33222-5G
76902-512EA 410.57 USD
76902-512 76902-514
2-Chloro-1-(methoxymethoxy)-3(trifluoromethyl)benzene ≥97%
2-Chloro-1-(methoxymethoxy)-3(trifluoromethyl)benzene
Formula: C₉H₈ClF₃O₂
MW: 240.607 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30723179
CAS Number: 1820814-26-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR