2-Chlor-6-methoxy-4-pyridineboronic acid pinacol ester ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
2-Chloro-6-methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 2-Chloro-6-methoxypyridine-4-boronic acid pinacol ester
33200-1G
33200-25G
33200-500MG
33200-5G
76902-496EA
290.69
USD
76902-496
76902-498
76902-500
76902-502
2-Chlor-6-methoxy-4-pyridineboronic acid pinacol ester ≥97%
2-Chlor-6-methoxy-4-pyridineboronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₂H₁₇BClNO₃ Storage Temperature: Refrigerator |
MDL Number:
MFCD11617855 CAS Number: 697739-24-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...