2-Isopropyl-4-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

33197-1G 33197-500MG 33197-5G
76902-484EA 498.63 USD
76902-484 76902-486 76902-488
2-Isopropyl-4-methoxybenzoic acid ≥97%
2-Isopropyl-4-methoxybenzoic acid
Formula: C₁₁H₁₄O₃
Storage Temperature: Refrigerator
MDL Number: MFCD14538440
CAS Number: 161480-96-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR