2-Hydroxy-3,5-dimethylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

33170-1G 33170-25G 33170-500MG 33170-5G
76902-470EA 123.07 USD
76902-470 76902-472 76902-474 76902-476
2-Hydroxy-3,5-dimethylbenzaldehyde ≥97%
2-Hydroxy-3,5-dimethylbenzaldehyde
Formula: C₉H₁₀O₂
Storage Temperature: Ambient
MDL Number: MFCD05625710
CAS Number: 24623-61-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR