2-Fluoro-5-formylphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Fluoro-5-formylbenzeneboronic acid pinacol ester, 4-Fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde, 2-Fluoro-5-formylphenylboronic acid, pinacol ester

33129-1G 33129-25G 33129-5G
76902-444EA 96.55 USD
76902-444 76902-446 76902-448
2-Fluoro-5-formylphenylboronic acid pinacol ester ≥97%
2-Fluoro-5-formylphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₆BFO₃
MW: 250.07 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11867877
CAS Number: 1112208-82-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR