2,6-Difluoro-3-methoxyphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

32860-25G 32860-5G
76902-422EA 360.71 USD
76902-422 76902-424
2,6-Difluoro-3-methoxyphenylboronic acid pinacol ester ≥97%
2,6-Difluoro-3-methoxyphenylboronic acid pinacol ester
Formula: C₁₃H₁₇BF₂O₃
MW: 270.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30529864
CAS Number: 2121514-34-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR