2,6-Difluoro-3-methoxybenzyl alcohol ≥95%

Supplier: AOBChem USA

Warning

32849-25G 32849-5G
76902-414EA 516.66 USD
76902-414 76902-416
2,6-Difluoro-3-methoxybenzyl alcohol ≥95%
2,6-Difluoro-3-methoxybenzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₈F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD04115904
CAS Number: 886498-45-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR