2-Hydroxymethylthiophen-3-boronic acid, pinacol ester ≥97%

Supplier: AOBChem USA

Warning

32687-1G 32687-25G 32687-500MG 32687-5G
76902-398EA 352.22 USD
76902-398 76902-400 76902-402 76902-404
2-Hydroxymethylthiophen-3-boronic acid, pinacol ester ≥97%
2-Hydroxymethylthiophen-3-boronic acid, pinacol ester
Formula: C₁₁H₁₇BO₃S
MW: 240.127 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD27923557
CAS Number: 2121511-82-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR