2-Methoxy-3-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

32669-25G 32669-5G
76902-390EA 595.17 USD
76902-390 76902-392
2-Methoxy-3-methylbenzaldehyde ≥97%
2-Methoxy-3-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₁₀O₂
MW: 150.175 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07783657
CAS Number: 67639-61-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR