2-Ethoxy-4-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

32558-25G 32558-5G
76902-380EA 289.63 USD
76902-380 76902-382
2-Ethoxy-4-methoxybenzaldehyde ≥97%
2-Ethoxy-4-methoxybenzaldehyde
Formula: C₁₀H₁₂O₃
MW: 180.2 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00016585
CAS Number: 42924-37-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR