2-Chloro-5-fluoro-3-(trifluoromethyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

32496-1G 32496-25G 32496-500MG 32496-5G
76902-342EA 225.98 USD
76902-342 76902-344 76902-346 76902-348
2-Chloro-5-fluoro-3-(trifluoromethyl)phenylboronic acid ≥97%
2-Chloro-5-fluoro-3-(trifluoromethyl)phenylboronic acid
Formula: C₇H₄BClF₄O₂
MW: 242.363 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD30529814
CAS Number: 2121513-70-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR