2-Bromo-1-ethoxy-3-fluorobenzene ≥95%

Supplier: AOBChem USA

Warning

32469-1G 32469-25G 32469-500MG 32469-5G
76902-328EA 284.33 USD
76902-328 76902-330 76902-332 76902-334
2-Bromo-1-ethoxy-3-fluorobenzene ≥95%
2-Bromo-1-ethoxy-3-fluorobenzene
Formula: C₈H₈BrFO
MW: 219.051 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD17215643
CAS Number: 1375098-48-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR