2,4-Dimethoxybenzoic acid ethyl ester ≥97%
Supplier: AOBChem USA
32463-25G
32463-5G
76902-316EA
310.85
USD
76902-316
76902-318
2,4-Dimethoxybenzoic acid ethyl ester ≥97%
2,4-Dimethoxybenzoic acid ethyl ester
Formula:
C₁₁H₁₄O₄ |
MDL Number:
MFCD02149437 CAS Number: 81722-07-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...